Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BHMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521466
Description
Recommended positive controls: HepG2, mouse liver. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (86%), Pig (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.
Specifications
BHMT | |
Polyclonal | |
Unconjugated | |
BHMT | |
betaine-homocysteine methyltransferase; betaine--homocysteine S-methyltransferase; betaine-homocysteine S-methyltransferase; betaine--homocysteine S-methyltransferase 1; BHMT; BHMT1; epididymis secretory sperm binding protein Li 61p; HEL-S-61p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12116, 635 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
O35490, Q93088 | |
BHMT | |
Recombinant protein encompassing a sequence within the center region of human BHMT. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction