Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BHMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114904
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114904 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114904 Supplier Invitrogen™ Supplier No. PA5114904
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BHMT.

This gene encodes a cytosolic enzyme that catalyzes the conversion of betaine and homocysteine to dimethylglycine and methionine, respectively. Defects in this gene could lead to hyperhomocyst(e)inemia, but such a defect has not yet been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHMT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene BHMT
Gene Accession No. O09171, O35490, Q93088
Gene Alias betaine-homocysteine methyltransferase; betaine--homocysteine S-methyltransferase; betaine-homocysteine S-methyltransferase; betaine--homocysteine S-methyltransferase 1; BHMT; BHMT1; epididymis secretory sperm binding protein Li 61p; HEL-S-61p
Gene Symbols BHMT
Host Species Rabbit
Immunogen A synthesized peptide derived from human BHMT(Accession Q93088), corresponding to amino acid residues D371-Q406.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12116, 635, 81508
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.