Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BHMT Rabbit anti-Human, Mouse, Rat, Clone: 7O7D7, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | BHMT |
---|---|
Clone | 7O7D7 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
BHMT Monoclonal antibody specifically detects BHMT in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
BHMT | |
Western Blot | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
635 | |
IgG | |
Affinity purified |
7O7D7 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
betaine homocysteine methyltransferase, betaine-homocysteine methyltransferase, betaine--homocysteine S-methyltransferase, betaine--homocysteine S-methyltransferase 1, BHMT1, EC 2.1.1, EC 2.1.1.5 | |
A synthetic peptide corresponding to a sequence within amino acids 300-406 of human BHMT (Q93088). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title