Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BHMT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560757 Supplier Invitrogen™ Supplier No. PA560757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTFSASEDNM ESKWEDVNAA ACDLAREVAG KGDALVAGGI CQTSIYKYQK DEA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

Homocysteine is a sulfur-containing amino acid that plays a crucial role in methylation reactions. Transfer of the methyl group from betaine to homocysteine creates methionine, which donates the methyl group to methylate DNA, proteins, lipids, and other intracellular metabolites. The protein encoded by this gene is one of two methyl transferases that can catalyze the transfer of the methyl group from betaine to homocysteine. Anomalies in homocysteine metabolism have been implicated in disorders ranging from vascular disease to neural tube birth defects such as spina bifida.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHMT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BHMT2
Gene Accession No. Q68FT5, Q91WS4, Q9H2M3
Gene Alias betaine-homocysteine methyltransferase 2; betaine--homocysteine S-methyltransferase 2; betaine-homocysteine S-methyltransferase 2; BHMT2; C81077; D13Ucla2; S-methylmethionine--homocysteine S-methyltransferase BHMT2; SMM-hcy methyltransferase
Gene Symbols BHMT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BHMT2. Recombinant protein control fragment (Product #RP-96851).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23743, 365972, 64918
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.