Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BHPI
GREENER_CHOICE

Catalog No. 611610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-161-0 10 mg
61-165-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-161-0 Supplier Tocris Bioscience™ Supplier No. 6116/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

ERα antagonist; also activates unfolded protein response; active in vivo

BHPI is an estrogen receptor (ER) α inhibitor. Inhibits proliferation of drug-resistant ERα-positive breast and ovarian cancer cells. Induces sustained activation of the unfolded protein response. Active in vivo.

Specifications

Chemical Name or Material 1,3-Dihydro-3,3-bis(4-hydroxyphenyl)-7-methyl-2H-indol-2-one
CAS 56632-39-4
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C21H17NO3
Formula Weight Observed MW: 331.36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.