Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHT, Granular, FCC, 99%, Spectrum™ Chemical
SDP

Catalog No. 18600977 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

B1095, 128-37-0, C15H24O

Spectrum™ Chemical BHT, Granular, FCC is used in small quantities as a food additive as an antioxidant. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 128-37-0
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.36
InChI Key NLZUEZXRPGMBCV-UHFFFAOYSA-N
IUPAC Name 2,6-di-tert-butyl-4-methylphenol
SMILES CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.002%
Packaging Fiber Drum
Quantity 12 kg
Percent Purity 99%
Grade FCC
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.