Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bi-1 Monoclonal Antibody (20F248), Invitrogen™

Catalog No. 01676623
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-623 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-623 Supplier Thermo Scientific Supplier No. MA141678

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

MA1-41678 is derived from a mixture of three synthetic peptides corresponding to the N-terminal, internal and C-terminal regions of human B1.  Epitope mapping indicates that the antibody recognizes an epitope in the N-terminal region of Bi-1.

BI-1 (Bax Inhibitor 1) is in anti-apoptotic protein that has been linked to protection from apoptosis induced endoplasmic reticulum (ER) stress (reviewed in Bailley-Maitre et al, 2007).  BI-1 contains several transmembrane domains, localizes to ER membranes, and has cytoprotective functions that are conserved in both animals and plants. BI-1 suppresses apoptosis induced by ectopic expression of the proapoptotic protein Bax as well as other types of stimuli.  Cells from BI-1 knockout (KO) mice are hypersensitivite to apoptosis induced by ER stress–causing chemical agents (thapsigargin, tunicamycin, and brefeldin A) or by ischemia-reperfusion (IR) injury.  Conversely, overexpression of BI-1 protects against apoptosis induced by ER stress and IR. The mechanism by which BI-1 protects cells from ER-stress induced apoptosis remains to be fully elucidated, it is thought to involve regulation of Ca2+ handling by the ER.   
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bi-1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 20F248
Concentration 1.0 mg/mL
Conjugate Unconjugated
Gene TMBIM6
Gene Accession No. P55061
Gene Alias BAXI1, BI-1, TEGT
Gene Symbols TMBIM6
Host Species Mouse
Immunogen A mixture of three synthetic peptides corresponding to the N-terminal, internal and C-terminal regions of human Bax inhibitor 1.
Purification Method IgM
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7009
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.