Learn More
Invitrogen™ Bi-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143940
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human RT4 whole cell, human CACO-2 whole cell. IHC: human endometrial adenocarcinoma tissue, human liver cancer tissue, human lung cancer tissue, human prostate cancer tissue. ICC/IF: Hela cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
BI-1 (Bax Inhibitor 1) is in anti-apoptotic protein that has been linked to protection from apoptosis induced endoplasmic reticulum (ER) stress (reviewed in Bailley-Maitre et al, 2007). BI-1 contains several transmembrane domains, localizes to ER membranes, and has cytoprotective functions that are conserved in both animals and plants. BI-1 suppresses apoptosis induced by ectopic expression of the proapoptotic protein Bax as well as other types of stimuli. Cells from BI-1 knockout (KO) mice are hypersensitivite to apoptosis induced by ER stress-causing chemical agents (thapsigargin, tunicamycin, and brefeldin A) or by ischemia-reperfusion (IR) injury. Conversely, overexpression of BI-1 protects against apoptosis induced by ER stress and IR. The mechanism by which BI-1 protects cells from ER-stress induced apoptosis remains to be fully elucidated, itis thought to involve regulation of Ca2+ handling by the ER.
Specifications
| Bi-1 | |
| Polyclonal | |
| Unconjugated | |
| TMBIM6 | |
| 5031406P05Rik; Ab1-011; Ac1-149; Bax inhibitor 1; Bax inhibitor-1; BAXI1; Bi1; BI-1; Cc1-27; Tegt; testis enhanced gene transcript; testis-enhanced gene transcript protein; Tmbim6; transmembrane BAX inhibitor motif containing 6; transmembrane BAX inhibitor motif-containing protein 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7009 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P55061 | |
| TMBIM6 | |
| A synthetic peptide corresponding to a sequence of human BI-1/TMBIM6 (RKINFDALLKFSHITPSTQQHLK). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.