Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BI 6015
GREENER_CHOICE

Catalog No. 464110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-411-0 10 mg
46-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-411-0 Supplier Tocris Bioscience™ Supplier No. 4641/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Hepatocyte nuclear factor 4α (HNF4α) antagonist

Hepatocyte nuclear factor 4α (HNF4α) antagonist. Represses expression of known HNF4α target genes. Decreases HNF4α DNA binding. Exhibits cytotoxic activity in a range of tumor cell lines, including human hepatocellular carcinoma.

Chemical Identifiers

CAS 93987-29-2
Molecular Formula C15H13N3O4S

Specifications

Quantity 10 mg
Formula Weight 331.35
Purity >98%
Molecular Weight (g/mol) 331.346
Chemical Name or Material BI 6015
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.