Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BI 9321
GREENER_CHOICESDP

Catalog No. 66655 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-655 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 66-655 Supplier R&D Systems™ Supplier No. 6665/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Nuclear receptor-binding SET domain (NSD) 3 antagonist; selectively binds PWWP1 domain

BI 9321 is a nuclear receptor-binding SET domain (NSD) 3 antagonist (IC50 = 1.2 μM). Selectively binds the PWWP1 domain of NSD3 (Kd = 166 nM). Antagonizes the interaction of H3 with NSD3-PWWP1 in U2OS cells.

Specifications

Chemical Name or Material

(4-(5-(7-Fluoroquinolin-4-yl)-1-methyl-1H-imidazol-4-yl)-3,5-dimethylphenyl)methanamine trihydrochloride

CAS 2387510-87-2
Quantity 5 mg
Solubility water to 100 mM; DMSO to 100 mM
Product Line Tocris
Molecular Formula C22H21FN4.3HCl
Formula Weight 469.81
Physical Form Solid
Color Yellow
Product Type Antagonist
Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.