Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bialaphos sodium, 85%

Catalog No. AAJ66802MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
Catalog No. Quantity
AAJ66802MC 100 mg
AAJ66802ME 500 mg
2 options

Catalog No. AAJ66802MC

Supplier: Thermo Scientific Chemicals J66802MC

Only null left
Add to Cart

Bialaphos sodium (bilanafos), CAS # 35597-43-4, is a tripeptide antibiotic used to confirm gene transfer in plants through use of the bar selection gene. | CAS: 71048-99-2 | C11H21N3NaO6P | 345.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

  • Bialaphos sodium consists of two L-alanyl units attached to L-phosphinothricyl
  • Bialaphos is a bacterial metabolite and an antimicrobial

Application

  • Bialaphos is commonly used for transformation experiments in some plant species as a selection agent
  • Bialaphos sodium can be used as a standard for metabolite and environmental contaminant determination in laboratory settings
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71048-99-2
Molecular Formula C11H21N3NaO6P
Molecular Weight (g/mol) 345.268
MDL Number MFCD01674432
InChI Key RTWIRLHWLMNVCC-WQYNNSOESA-M
PubChem CID 118988468
IUPAC Name sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate
SMILES CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+]

Specifications

Type Bialaphos Sodium
Physical Form Powder
Melting Point 160°C
Color Orange
Quantity 100 mg
Flash Point >110°C (230°F)
UN Number UN2811
Boiling Point 793°C
Solubility Information Soluble in water
Formula Weight 345.26
Percent Purity 85%
Chemical Name or Material Bialaphos sodium
Show More Show Less
Hazard Category H301-H315-H319
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber AY6827000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.