Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bianthrone 95.0+%, TCI America™

Click to view available options
Quantity:
100 mg
1 g
5 g
Specifications
Specifications
Melting Point | 300°C |
Color | Green-Yellow |
Quantity | 1 g |
Formula Weight | 384.43 |
Percent Purity | ≥95.0% (HPLC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Bianthrone |
Safety and Handling
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction