Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BIBR 1532
GREENER_CHOICE

Catalog No. 298110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-811-0 10 mg
29-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-811-0 Supplier Tocris Bioscience™ Supplier No. 2981/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective telomerase inhibitor

Selective telomerase inhibitor (IC50 values are 93, > 100000 and > 100000 nM for human telomerase, human RNA polymerase I and human RNA polymerase II + III respectively). Causes telomere shortening in exponentially growing NCI-H460 lung carcinoma cells and eventual growth arrest.

Chemical Identifiers

CAS 321674-73-1
Molecular Formula C21H17NO3

Specifications

Quantity 10 mg
Formula Weight 331.36
Purity >98%
Molecular Weight (g/mol) 331.371
Chemical Name or Material BIBR 1532
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.