Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ BICC1/Protein Bicaudal C Homolog 1, Mouse, Unlabeled, Clone: 2D9.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MABE344
Description
Protein bicaudal C homolog 1 (UniProt Q9H694; also known as Bic-C) is encoded by the BICC1 (also known as CYSRD) gene (Gene ID 80114) in human. Drosophila bicaudal C and its mammalian homologs are RNA-binding proteins involved in symmetric distribution and localized translation of mRNAs of many proteins in a wide range of cells and tissues. RNA binding of Bic-C is mediated by N-terminal KH domains (a.a. 132-199 and 284-348), whereas a C-terminal sterile alpha motif (SAM; a.a. 873-936) self-polymerizes and localizes Bic-C at cytoplasmic foci where silencing of bound mRNAs takes place. Defective polymerization results in decreased Bic-C stability and thereby attenuates inhibition of Dishevelled-2 of Wnt/β-catenin pathway. Loss of BICC1 provokes renal and pancreatic cysts as well as ectopic Wnt/β-catenin signaling during visceral left-right patterning due to defective silencing of Bic-C target mRNAs, including adenylate cyclase 6 (AC6).
Specifications
BICC1/Protein Bicaudal C Homolog 1 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide. | |
Q9H694 | |
Recombinant fragment corresponding to the N-terminal half of human BICC1/Protein Bicaudal C Homolog 1. | |
100 μg | |
Epigenetics & Nuclear Function | |
NP_001073981 | |
Human | |
Purified |
Western Blot | |
2D9.1 | |
Unconjugated | |
BICC1, CYSRD | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 2D9.1 reacts with both spliced isoforms of human and murine BICC1 reported by UniProt (Q9H694 & Q99MQ1). | |
Stable for 1 year at 2°-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction