Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BICD2 Monoclonal Antibody (GT1824)
GREENER_CHOICE

Catalog No. PIMA527825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527825 Supplier Invitrogen™ Supplier No. MA527825
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, PC-12, Rat2 Predicted Reactivity: Mouse (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BICD2 is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BICD2
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GT1824
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene BICD2
Gene Accession No. Q8TD16
Gene Alias bA526D8.1; bicaudal D homolog 2; bicaudal D homolog 2 (Drosophila); bicaudal D protein; Bic-D 2; BICD cargo adaptor 2; BICD2; coiled-coil protein BICD2; cytoskeleton-like bicaudal D protein homolog 2; homolog of Drosophila bicaudal D; KIAA0699; Protein bicaudal D homolog 2; RP11-476B13.3; SMALED2
Gene Symbols BICD2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human BICD2. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23299, 306809
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.