Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BICD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582810 Supplier Invitrogen™ Supplier No. PA582810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSAPSEEEEY ARLVMEAQPE WLRAEVKRLS HELAETTREK IQAAEYGLAV LEEKHQLKLQ FEELEVDYEA I Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

BICD2 is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BICD2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BICD2
Gene Accession No. Q8TD16, Q921C5
Gene Alias bA526D8.1; bicaudal D homolog 2; bicaudal D homolog 2 (Drosophila); bicaudal D protein; Bic-D 2; BICD cargo adaptor 2; BICD2; coiled-coil protein BICD2; cytoskeleton-like bicaudal D protein homolog 2; homolog of Drosophila bicaudal D; KIAA0699; Protein bicaudal D homolog 2; RP11-476B13.3; SMALED2
Gene Symbols BICD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BICD2. Recombinant protein control fragment (Product #RP-93077).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23299, 306809, 76895
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.