Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BICD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587781 Supplier Invitrogen™ Supplier No. PA587781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVAVAEAGEL REQLKALRST HEAREAQHAE EKGRYEAEGQ ALTEKVSLLE KASRQDRELL ARLEKELKKV SDVAGETQGS LSVAQDELVT FSEELANLYH HVCMCNNETP NRVMLDYYRE GQGGAGRTSP GGRTSPEARG RRSPILLPKG LLAPEAGRAD GGTGDSSPSP GSSLPSPLSD P; Positive Samples: U-87MG, HL-60, HeLa, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Golgi apparatus, cytoskeleton.

BICD2 is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BICD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BICD2
Gene Accession No. Q8TD16, Q921C5
Gene Alias bA526D8.1; bicaudal D homolog 2; bicaudal D homolog 2 (Drosophila); bicaudal D protein; Bic-D 2; BICD cargo adaptor 2; BICD2; coiled-coil protein BICD2; cytoskeleton-like bicaudal D protein homolog 2; homolog of Drosophila bicaudal D; KIAA0699; Protein bicaudal D homolog 2; RP11-476B13.3; SMALED2
Gene Symbols BICD2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 440-620 of human BICD2 (NP_0010038001).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23299, 306809, 76895
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.