Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BICD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598718
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598718 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598718 Supplier Invitrogen™ Supplier No. PA598718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BICD2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

BICD2 is one of two human homologs of Drosophila bicaudal-D and a member of the Bicoid family. It has been implicated in dynein-mediated, minus end-directed motility along microtubules. It has also been reported to be a phosphorylation target of NIMA related kinase 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BICD2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.945 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene BICD2
Gene Accession No. Q8TD16
Gene Alias bA526D8.1; bicaudal D homolog 2; bicaudal D homolog 2 (Drosophila); bicaudal D protein; Bic-D 2; BICD cargo adaptor 2; BICD2; coiled-coil protein BICD2; cytoskeleton-like bicaudal D protein homolog 2; homolog of Drosophila bicaudal D; KIAA0699; Protein bicaudal D homolog 2; RP11-476B13.3; SMALED2
Gene Symbols BICD2
Host Species Rabbit
Immunogen Recombinant Human Protein bicaudal D homolog 2 protein (511-625AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23299
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.