Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicine, 99.38%, MP Biomedicals™

Catalog No. ICN10100525
Click to view available options
Quantity:
25 g
100 g
250 g

Chemical Identifiers

CAS 150-25-4
Molecular Formula C6H12NNaO4
Molecular Weight (g/mol) 185.16
MDL Number MFCD00004295
InChI Key MFBDBXAVPLFMNJ-UHFFFAOYSA-M
Synonym N, N-bis [2-hydroxyethyl]glycine
PubChem CID 8761
ChEBI CHEBI:40957
SMILES [Na+].OCCN(CCO)CC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 0.9938
IUPAC Name sodium 2-[bis(2-hydroxyethyl)amino]acetate
Formula Weight 163.2
Chemical Name or Material Bicine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.