Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (-)-Bicuculline methiodide
GREENER_CHOICESDP

Catalog No. 250310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-031-0 10 mg
25-035-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-031-0 Supplier Tocris Bioscience™ Supplier No. 2503/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

GABAA antagonist; more water soluble version of (+)-bicuculline (Cat. No. 0130)

Methiodide form of classical GABAA receptor antagonist (+)-bicuculline . More water-soluble and stable. Non-GABA receptor-mediated actions reported, including actions on calcium-dependent potassium channels.

Methobromide Salt and Methochloride Salt also available.

Chemical Identifiers

CAS 40709-69-1
Molecular Formula C21H20INO6

Specifications

Quantity 10 mg
Formula Weight 509.3
Purity >99%
Molecular Weight (g/mol) 509.296
Chemical Name or Material (-)-Bicuculline methiodide
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.