Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Bicuculline 98.0+%, TCI America™
SDP

Catalog No. B1890100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1890100MG 100 mg
B189025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1890100MG Supplier TCI America Supplier No. B1890100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 485-49-4
Molecular Formula C20H17NO6
Molecular Weight (g/mol) 367.357
MDL Number MFCD00005006
InChI Key IYGYMKDQCDOMRE-ZWKOTPCHSA-N
Synonym +-bicuculline, bicuculline, d-bicuculline, bicculine, bicucullin, bicuculline +, unii-y37615dvkc, 6r-6-5s-6-methyl-5,6,7,8-tetrahydro 1,3 dioxolo 4,5-g isoquinolin-5-yl furo 3,4-e 1,3 benzodioxol-8 6h-one, furo 3,4-e-1,3-benzodioxol-8 6h-one, 6-5,6,7,8-tetrahydro-6-methyl-1,3-dioxolo 4,5-g isoquinolin-5-yl-, r-r*,s*, 6r-6-5s-6-methyl 5,6,7,8-tetrahydro-2h-1,3-dioxolano 4,5-g isoquinolin-5-yl-6-hydro-2h-1,3-dioxoleno 4,5-e isobenzofuran-8-one
PubChem CID 10237
ChEBI CHEBI:3092
IUPAC Name (6R)-6-[(5S)-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-6H-furo[3,4-g][1,3]benzodioxol-8-one
SMILES CN1CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3

Specifications

Melting Point 197°C
Color Yellow
Quantity 100 mg
UN Number 1544
Formula Weight 367.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Bicuculline
RTECSNumber LV0909840
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.