Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[3.2.1]oct-2-ene 98.0+%, TCI America™
SDP

Catalog No. B19141G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 823-02-9
Molecular Formula C8H12
Molecular Weight (g/mol) 108.184
MDL Number MFCD00078236
InChI Key YEHSTKKZWWSIMD-UHFFFAOYSA-N
PubChem CID 136649
IUPAC Name bicyclo[3.2.1]oct-3-ene
SMILES C1CC2CC1CC=C2

Specifications

Melting Point 39°C
Color White
Boiling Point 135°C
Quantity 1 g
Formula Weight 108.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Bicyclo[3.2.1]oct-2-ene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.