Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BID Antibody (002), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289515G
Description
BID Monoclonal specifically detects BID in Human samples. It is validated for Western Blot, ELISA.Specifications
BID | |
Monoclonal | |
DyLight 488 | |
apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human BID (P55957-1; Met1-Asp195). | |
0.1 mL | |
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors | |
637 | |
Human | |
Purified |
Western Blot, ELISA | |
2 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Has cross-reactivity in ELISA with: Mouse BID | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction