Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BID Rabbit anti-Human, Unconjugated, Clone: AAGC-2, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19622
Description
BID Monoclonal antibody specifically detects BID in Human samples. It is validated for IP, ICC, WB-Ce, IHC-P, IF, Flow Cyt.Specifications
BID | |
Monoclonal | |
Unconjugated | |
BID | |
Rabbit | |
100 μg | |
637 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
AAGC-2 | |
BH3 interacting domain death agonist | |
FP497, MGC15319, MGC42355 | |
A synthetic peptide corresponding to human BID | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction