Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biebrich Scarlet sodium salt

Catalog No. p-7043119
Click to view available options
Quantity:
25 g
100 g

Catalog No. AAJ6560314

Supplier: Thermo Scientific Chemicals J6560314

Only null left
Add to Cart

CAS: 4196-99-0 | C22H14N4Na2O7S2 | 556.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4196-99-0
Molecular Formula C22H14N4Na2O7S2
Molecular Weight (g/mol) 556.47
MDL Number MFCD00003891
InChI Key YCKMEQRBEVVZQF-VKZXTWAVSA-L
Synonym C.I. 26905; Acid Red 66
PubChem CID 16219040
IUPAC Name disodium 2-{2-[(1Z)-2-oxo-1,2-dihydronaphthalen-1-ylidene]hydrazin-1-yl}-5-[2-(4-sulfonatophenyl)diazen-1-yl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(N\N=C2/C(=O)C=CC3=CC=CC=C23)C(=C1)S([O-])(=O)=O

Specifications

Melting Point 181°C to 188°C
Beilstein 3883900
Formula Weight 556.49
Color Brown to Red
Quantity 25 g
Flash Point 300°C (572°F)
Physical Form Powder
Chemical Name or Material Biebrich Scarlet sodium salt
EINECSNumber 224-084-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.