Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bif1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554111
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554111 Supplier Invitrogen™ Supplier No. PA554111
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: THAHHLRCLN DFVEAQMTYY AQCYQYMLDL QKQLGSFPSN YLSNNNQTSV TPVPSVLPNA IGSSAMASTS GLVITSPSNL SDLKECSGSR KARVLYDYDA ANSTELSLLA DEVITVFSVV GMDSD Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

Bif1 gene encodes a protein similar to a protein in rodents which is induced by bone morphogenic protein 2 in vitro and may be involved in BMP2-induced transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bif1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Sh3glb1
Gene Accession No. Q6AYE2, Q9Y371
Gene Alias AA409932; AI314629; AU015566; Bax-interacting factor 1; BIF1; Bif-1; CGI-61; dJ612B15.2; EMBL:AAH79085.1}; endophilin b1; endophilin-B1; Kiaa0491; mKIAA0491; PPP1R70; protein phosphatase 1, regulatory subunit 70; SH3 domain containing GRB2 like endophilin B1; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like B1 (endophilin); SH3-domain GRB2-like endophilin B1; SH3GLB1; sh3glb1 {ECO:0000312; testicular tissue protein Li 172
Gene Symbols Sh3glb1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Bif1. Recombinant protein control fragment (Product #RP-90707).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292156, 51100
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.