Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bifonazole 98.0+%, TCI America™
SDP

Catalog No. B417325G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 60628-96-8
Molecular Formula C22H18N2
Molecular Weight (g/mol) 310.4
MDL Number MFCD00865567
InChI Key OCAPBUJLXMYKEJ-UHFFFAOYSA-N
Synonym bifonazole, mycospor, trifonazole, bifonazol, bifonazolum, amycor, azolmen, bifonazol inn-spanish, bifonazolum inn-latin, 1-4-biphenylyl phenylmethyl-1h-imidazole
PubChem CID 2378
ChEBI CHEBI:78692
IUPAC Name 1-[phenyl-(4-phenylphenyl)methyl]imidazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 25 g
Formula Weight 310.40
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Bifonazole
RTECSNumber NI3517000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.