Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bifonazole, 99%

Catalog No. p-7042654
Click to view available options
Quantity:
5 g

CAS: 60628-96-8 | C22H18N2 | 310.4 g/mol

Bifonazole is an imidazole antifungal agent and calmodulin antagonist. It causes a reduction in glycolysis and ATP levels in B16 melanoma cells. Bifonazole is an inhibitor of C14orf1. Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bifonazole is an imidazole antifungal agent and calmodulin antagonist. It causes a reduction in glycolysis and ATP levels in B16 melanoma cells. Bifonazole is an inhibitor of C14orf1. Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.

Solubility
Soluble in DMSO

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60628-96-8
Molecular Formula C22H18N2
Molecular Weight (g/mol) 310.4
MDL Number MFCD00865567
InChI Key OCAPBUJLXMYKEJ-UHFFFAOYSA-N
Synonym Mycosporan; 1-(alpha,4-Diphenylbenzyl)imidazole
PubChem CID 2378
ChEBI CHEBI:78692
IUPAC Name 1-[phenyl-(4-phenylphenyl)methyl]imidazole
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4

Specifications

Type Bifonazole
Physical Form Powder
Melting Point 142°C
Color White
Quantity 5 g
Merck Index 14,1213
Solubility Information Soluble in DMSO
Formula Weight 310.39
Percent Purity 99%
Chemical Name or Material Bifonazole
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 262-336-6
RTECSNumber NI3517000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.