Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Big ET-1 ELISA Kit

Catalog No. p-200020541
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 Tests
96 Tests

ELISA

The protein encoded by this gene is proteolytically processed to release a secreted peptide termed endothelin 1. This peptide is a potent vasoconstrictor and is produced by vascular endothelial cells. Endothelin 1 also can affect the central nervous system. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05305
Assay Range 0.4 to 25 pg/mL
Assay Sensitivity 0.185 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates, Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1906
Gene Symbol EDN1
Interassay CV 4.9%
Intraassay CV 4.7
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, Streptavidin-HRP, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL
Storage Requirements 2°C to 8°C
Target Endothelin 1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.