Description
- High capacity filters ideal for bulk purification - cumulative capacity level of 100μg/L of oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide at a flow rate of 8L/min
- Efficiency < 100 ppb
- Can be used where several instruments are plumbed from a single gas source
- Sintered stainless steel frits prevent particulate contamination
Specifications
Specifications
| For Use With (Application) | GC and GCMS applications where helium is employed |
| Model | Hydrocarbon |
| Volume (Metric) | 750 cm3 |
| Port Type | 1/4 in. |
| Description | Removes oxygen, moisture, hydrocarbons, carbon dioxide and carbon monoxide |
| For Use With | Big Trap Mounting Clip |
| Pressure | 250 psig |