Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIG1 Mouse anti-Human, Clone: 102, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS1247
Description
Detect BIG1 using this mouse monoclonal Anti-BIG1 Antibody, clone 102. It has been tested in Immunocytochemistry and Western Blotting.
Specifications
BIG1 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
ARFGEF1,ARFGEP1,BIG1 | |
GST-tagged recombinat fragment corresponding to 311 amino acids from human Brefeldin A-inhibited guanine nucleotide-exchange protein 1 (BIG1). | |
100 μg | |
Signaling | |
NP_006412 | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
102 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 102 specifically detects BIG1 protein in human and targets an epitope within amino acids 212-522 in the N-terminal region. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction