Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIG2 Mouse anti-Human, Clone: 56, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS1246
Description
Detect BIG2 using this mouse monoclonal Anti-BIG2 Antibody, clone 56 Immunoprecipitation, and Western Blotting.
Specifications
BIG2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
ARFGEF2,ARFGEP2,BIG2 | |
GST-tagged recombinant fragment corresponding to 249 amino acids from the N-terminal region of human Brefeldin A-inhibited guanine nucleotide-exchange protein 2 (BIG2). | |
100 μg | |
Signaling | |
NP_006411 | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
56 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 56 specifically detects BIG2 in human. It targets an epitope within amino acids 9-257 in the N-terminal region. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction