Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer

Catalog No. p-7138582
Encompass_Preferred
Click to view available options
Quantity:
1 mL
28 mL
233 mL

This sequencing buffer is optimized for use with BigDye™ Terminator v1.1 and v3.1 Cycle Sequencing Kits.

  • Optimized for use with BigDye™ Terminator Cycle Sequencing Kits

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Buffer Sequencing Buffer
Concentration 5X
Content And Storage Store at 2°C to 8°C.
For Use With (Equipment) GeneAmp 9700, Veriti Thermal Cycler, 3500 Genetic Analyzer, 3730xl DNA Analyzer, 3130xl Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer
For Use With (Application) Standard Sequencing
Product Line BigDye™
Product Type Sequencing Buffer
Quantity 1 mL
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.