Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Biglycan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596232 Supplier Invitrogen™ Supplier No. PA596232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQRGFWDFTL DDGPFMMNDE EASGADTSGV LDPDSVTPTY SAMCPFGCHC HLRVVQCSDL GLKSVPKEIS PDTTLLDLQN NDISELRKDD FKGLQHLYAL VLVNNKISKI HEKAFSPLRK LQKLYISKNH LVEIPPNLPS SLVELRIHDN RIRKVPKGVF SGLRNMNCIE MGGNPLENSG FEPGAFDGLK LNYLRISEAK LTGIPKDLPE T; Positive Samples: U-87MG, HepG2; Cellular Location: extracellular matrix, extracellular space.

BGN is a small cellular or pericellular matrix proteoglycan that is closely related in structure to two other small proteoglycans, decorin and fibromodulin. The protein and decorin are thought to be the result of a gene duplication. Decorin contains one attached glycosaminoglycan chain, while this protein probably contains two chains. For this reason, this protein is called biglycan. This protein is thought to function in connective tissue metabolism by binding to collagen fibrils and transferring growth factor-beta. It may promote neuronal survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biglycan
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BGN
Gene Accession No. P21810
Gene Alias BG; BGN; Biglycan; bone/cartilage proteclycan 1; bone/cartilage proteoglycan I; bone/cartilage proteoglycan-I; BSPG1; dermatan sulphate proteoglycan I; DSPG1; PG S1; PGI; PG-S1; SLRR1A; small leucine-rich protein 1A; Small proteoglycan I (biglycan), bone (BSPG1) (bone/cartilage proteclycan 1 precursor)
Gene Symbols BGN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-230 of human BGN (NP_0017021).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 633
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.