Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Biglycan Recombinant Rabbit Monoclonal Antibody (JB71-31)
GREENER_CHOICE

Catalog No. PIMA534686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534686 Supplier Invitrogen™ Supplier No. MA534686
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: HepG2, human kidney tissue, mouse heart tissue, human skin tissue lysates.

BGN is a small cellular or pericellular matrix proteoglycan that is closely related in structure to two other small proteoglycans, decorin and fibromodulin. The protein and decorin are thought to be the result of a gene duplication. Decorin contains one attached glycosaminoglycan chain, while this protein probably contains two chains. For this reason, this protein is called biglycan. This protein is thought to function in connective tissue metabolism by binding to collagen fibrils and transferring growth factor-beta. It may promote neuronal survival.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Biglycan
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JB71-31
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene BGN
Gene Accession No. P21810
Gene Alias BG; BGN; Biglycan; bone/cartilage proteclycan 1; bone/cartilage proteoglycan I; bone/cartilage proteoglycan-I; BSPG1; dermatan sulphate proteoglycan I; DSPG1; PG S1; PGI; PG-S1; SLRR1A; small leucine-rich protein 1A; Small proteoglycan I (biglycan), bone (BSPG1) (bone/cartilage proteclycan 1 precursor)
Gene Symbols BGN
Host Species Rabbit
Immunogen Recombinant protein within Human Biglycan aa 260-368.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 633
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.