Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIK Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298984MFV500
Description
BIK Polyclonal antibody specifically detects BIK in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
BIK | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
Apoptosis inducer NBK, bcl-2-interacting killer, BCL2-interacting killer (apoptosis-inducing), BIP1, BP4, NBKapoptosis-inducing NBK | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human BIK. | |
0.1 mL | |
Apoptosis, Phospho Specific | |
638 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction