Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bik/NBK Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989932
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9932 Supplier Bioss Supplier No. BS23762R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Bik protein is known to interact with cellular and viral survival-promoting proteins, such as BCL2 and the Epstein-Barr virus in order to enhance programmed cell death. Because its activity is suppressed in the presence of survival-promoting proteins, this protein is suggested as a likely target for antiapoptotic proteins. This protein shares a critical BH3 domain with other death-promoting proteins, BAX and BAK.

Specifications

Antigen Bik/NBK
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene BIK
Gene Accession No. Q13323
Gene Alias Apoptosis inducer NBK; apoptosis-inducing NBK; BBC1; BCL2 interacting killer; Bcl-2-interacting killer; BCL2-interacting killer; BCL2-interacting killer (apoptosis-inducing); bhikhari; Bik; Bik-like killer protein; Biklk; BIP 1; BIP1; Blk; BP 4; BP4; cb 60; natural born killer; Nbk
Gene Symbols BIK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Bik/NBK.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 638
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.