Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BIKE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555322
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555322 Supplier Invitrogen™ Supplier No. PA555322
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YSANRSVADK EAIANFTNQK NISNPPDMSG WNPFGEDNFS KLTEEELLDR EFDLLRSNRL EERASSDKNV DSLSAPHNH Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.

The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. BMP2K (BMP2 inducible kinase), also known as BIKE, is a 1,161 amino acid nuclear protein that contains one protein kinase domain and belongs to the Ser/Thr protein kinase family. Thought to be involved in osteoblast differentiation, BMP2K catalyzes the ATP-dependent phosphorylation of bone morphogenic proteins (BMPs); proteins that are essential for proper cartilage and bone formation. Via its catalytic activity, BMP2K may play a role in signaling pathways that mediate bone growth and cellular differentiation. Three isoforms of BMP2K exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIKE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BMP2K
Gene Accession No. Q9NSY1
Gene Alias 4933417M22Rik; AA673486; AV128808; BIKE; Bike kinase; BMP2 inducible kinase; BMP-2 inducible kinase; BMP-2-inducible protein kinase; Bmp2k; HRIHFB2017
Gene Symbols BMP2K
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BIKE. Recombinant protein control fragment (Product #RP-95068).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55589
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.