Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bile Acid Receptor NR1H4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982429
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2429 Supplier Bioss Supplier No. BS12867R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The Vasoactive Intestinal Polypeptide Receptor 2 (VPAC2) is a member of a family of three G-protein coupled receptors which mediate VIP (vasoactive intestinal peptide) and PACAP (pituitary adenlyate cyclase activating peptide) function in a variety of tissues. Together, the VPAC1 and VPAC2 receptors compose a subtype of the VIP receptor. Expression of the VIP and PACAP receptors has been linked to the presence of human tumors.

Specifications

Antigen Bile Acid Receptor NR1H4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NR1H4
Gene Accession No. Q96RI1
Gene Alias AI957360; BAR; Bile acid receptor; BOS_5275; farnesoid X activated receptor; farnesoid X nuclear receptor; Farnesoid X-activated receptor; Farnesol receptor HRR-1; FXR; HRR1; HRR-1; MGC163445; Nr1h4; Nuclear receptor subfamily 1 group H member 4; nuclear receptor subfamily 1, group H, member 4; Retinoid X receptor-interacting protein 14; RIP14; RXR-interacting protein 14; Rxrip14
Gene Symbols NR1H4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Bile Acid Receptor NR1H4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9971
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.