Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Bile Esculin Agar
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
15mm x 103mm Tube - Slant
Monoplate
Media Type:
Bile Esculin Agar
Manufacturing Origin:
United States
Description
Isolate and differentiate enterococci/Group D streptococci and non Group D streptococci using Thermo Scientific™ Bile Esculin Agar, a selective microbiological medium.
Enterococci / Group D streptococci hydrolyse aesculin to form aesculetin and dextrose. Aesculetin combines with ferric citrate in the medium to form a dark brown or black complex which is indicative of a positive result. Bile salts will inhibit Gram-positive bacteria other than enterococci / Group D streptococci.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.

Specifications
Specifications
Format | Monoplate |
Manufacturing Origin | United States |
Media Type | Bile Esculin Agar |
Product Type | Agar |
Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction