Learn More
Thermo Scientific™ Bile Esculin Azide Agar Available on GSA/VA Contract for Federal Government customers only.
Isolate, enumerate and presumptively identify group D streptococci and enterococci with a medium that utilizes bile salts to improve selectivity.
$72.35 - $993.83
Specifications
Description Bile Esculin Azide Agar
View More Specs
Products 4
Catalog Number Mfr. No. Description Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Format Quantity Price Quantity & Availability  
R01189 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R01189
Bile Esculin Azide Agar 12mm x 85mm Monoplate 100/Pk. Case of 100 for $993.83
Add to cart
R01194 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R01194
Bile Esculin Azide Agar 12mm x 85mm Monoplate 10/Pk. Pack of 10 for $72.35
Add to cart
R01195 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R01195
Bile Esculin Azide Agar 10mm x 100mm Spacesaver Plate 15/Pk. Pack of 15 for $134.51
Add to cart
R060298
View Documents
Thermo Scientific™
R060298
Bile Esculin Azide Agar 15mm x 103mm Tube - Slant 20/Pk. Pack of 20 for $197.82
Add to cart
Description
Description
Isolate, enumerate and presumptively identify group D streptococci and enterococci using Thermo Scientific™ Bile Esculin Azide Agar, a microbiological media that includes a high portion of Ox bile.
Ox bile present in the media inhibits the growth of Gram-positive bacteria except Enterococci, while Sodium azide suppresses the Gram-negative bacteria. Group D streptococci and enterococci are able to hydrolyse aesculin into aesculetin and glucose, and the bondage of aesculetin and ferric ammonium citrate causes the brown-black to black halo around the colonies.
Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Bile Esculin Azide Agar