Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bile salt-activated lipase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986292
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6292 Supplier Bioss Supplier No. BS5042R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Upon antigen challenge, T-helper cells differentiate into two functional distinct subsets, Th1 and Th2. Th1 cells produce IL-2, IFN-gamma and lymphotoxin-beta that augment cell mediated immune response while Th2 cells secrete IL-4, IL-5, and IL-10 that enhance humoral immunity. The function of T-helper cells is regulated by cytokines. A novel cytokine receptor was recently identified and cloned. It is a new member in the type I cytokine receptor family and designated TCCR for T-cell cytokine receptor and WSX-1. TCCR deficient mice had impaired Th1 responses to protein antigen challenge, including decreased levels of IFN-gamma and Th1-dependent antibody IgG2a. TCCR is predominately expressed in thymus, spleen, lymph notes and peripheral blood leukocytes.

Specifications

Antigen Bile salt-activated lipase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CEL
Gene Accession No. P19835
Gene Alias 1810036E18Rik; BAL; Bile salt-activated lipase; bile salt-dependent lipase, oncofetal isoform; Bile salt-stimulated lipase; BSDL; BSSL; bucelipase; carboxyl ester hydrolase; Carboxyl ester lipase; carboxyl ester lipase (bile salt-stimulated lipase); CEase; Cel; CELL; Cholesterol esterase; FAP; FAPP; fetoacinar pancreatic protein; Lip1; LIPA; lysophospholipase, pancreatic; MODY8; Pancreatic lysophospholipase; Sterol esterase
Gene Symbols CEL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Bile salt-activated lipase .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1056
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.