Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bilirubin, 98+%

Catalog No. AC230225000
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle

Bilirubin, CAS # 635-65-4, is a tetrapyrroliccompound found in bile that is a product of heme catabolism. | CAS: 635-65-4 | C33H36N4O6 | 584.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bilirubin is a degradation product of heme following the activity of heme oxidase. Bilirubin is produced in the reticuloendothelial system and conjugated in the liver. It appears as an orange-yellow pigment that is secreted in the bile
  • High levels of unconjugated bilirubin can result in jaundice, a condition of the yellowing of skin and eyes. This can be toxic and may be a sign of severe liver disease
  • Bilirubin has been seen to have functions including cell signaling, metabolism modulation, and immune regulation
  • It has protective antioxidant activity at moderate levels

Application

  • Bilirubin is widely used in studies of liver function and disease, such as hepatitis or cirrhosis
  • As an antioxidant, bilirubin has been studied in protection against H2O2-mediated cell death
  • It can be used as a scavenger of reactive oxygen species (ROS) or as an inhibitor of NADPH oxidase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 635-65-4
Molecular Formula C33H36N4O6
Molecular Weight (g/mol) 584.67
MDL Number MFCD00005499
InChI Key BPYKTIZUTYGOLE-IFADSCNNSA-N
Synonym bilirubin, hematoidin, hemetoidin, bilirubin ix-alpha, principal bile pigment, unii-rfm9x3lj49, bilirubin ixalpha, 21h-biline-8,12-dipropanoic acid, 2,17-diethenyl-1,10,19,22,23,24-hexahydro-3,7,13,18-tetramethyl-1,19-dioxo, rfm9x3lj49, biline-8,12-dipropionic acid, 1,10,19,22,23,24-hexahydro-2,7,13,17-tetramethyl-1,19-dioxo-3,18-divinyl
PubChem CID 5280352
ChEBI CHEBI:16990
IUPAC Name 3-[2-[[3-(2-carboxyethyl)-5-[(Z)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methyl]-5-[(Z)-(4-ethenyl-3-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-3-yl]propanoic acid
SMILES CC1=C(C=C)\C(NC1=O)=C\C1=C(C)C(CCC(O)=O)=C(CC2=C(CCC(O)=O)C(C)=C(N2)\C=C2/NC(=O)C(C=C)=C2C)N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown-Red to Orange
Loss on Drying 1.8% max. (60°C, 2 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 500 mg
Merck Index 15, 1222
Formula Weight 584.67
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Bilirubin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.