Learn More
Specifications
Specifications
| For Use With (Application) | SDS-PAGE, Western Blot |
| Formulation | 10mM Tris pH7.5, 0.1mM EDTA, 0.2mM DTT, 20% glycerol |
| Gene ID (Entrez) | 644 |
| Molecular Weight (g/mol) | M.W. Theoretical: 36 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -70C. Avoid freeze-thaw cycles. |
| Gene Alias | biliverdin reductase A, Biliverdin-IX alpha-reductase, BLVR, BVR, BVR A, BVRA, EC 1.3.1.24 |
| Gene Symbol | BLVRA |
| Research Category | Cellular Signaling, metabolism, Protein Phosphatase, Serine Kinases |
| Purity or Quality Grade | >85% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
