Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bim Antibody (1C2H4), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP175511AF488
Description
Bim Monoclonal specifically detects Bim in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Bim | |
Monoclonal | |
Alexa Fluor 488 | |
O43521 | |
BCL2L11 | |
Antibody was raised against a recombinant protein corresponding to amino acids 2 to 198 of human Bim. | |
0.1 mL | |
Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific, Tumor Suppressors | |
10018 | |
Store at 4C in the dark. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1C2H4 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction