Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bim Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $493.10

Specifications

Antigen Bim
Concentration 0.6 mg/mL
Dilution Western Blot 2.0 ug/ml, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:100
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP253088SS
SDP
View Documents
Novus Biologicals
NBP253088SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP253088
SDP
View Documents
Novus Biologicals
NBP253088
0.1 mg
Each for $493.10
Only null left
Add to Cart
 
Description

Description

Bim Polyclonal specifically detects Bim in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Bim
Western Blot 2.0 ug/ml, Immunohistochemistry 1:50 - 1:100, Immunocytochemistry/Immunofluorescence 10-20 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:100
Polyclonal
Rabbit
Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific, Tumor Suppressors
PBS with 1% BSA with 0.02% Sodium Azide
BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL
BCL2L11
IgG
Affinity Purified
0.6 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse
O43521
10018
Partial recombinant human Bim protein (amino acids 2-120) UniProt O43521]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.