Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bim Rabbit anti-Human, Mouse, Rat, Clone: 3R0T2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Bim |
---|---|
Clone | 3R0T2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Bim Monoclonal antibody specifically detects Bim in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Bim | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
10018 | |
IgG | |
Affinity purified |
3R0T2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Bim (O43521). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title