Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BIMU 8
GREENER_CHOICE

Catalog No. 437425
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-742-5 25 mg
43-745 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-742-5 Supplier Tocris Bioscience™ Supplier No. 4374/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent 5-HT4 agonist

BIMU 8 is a potent 5-HT4 receptor full agonist (KD = 77 nM and EC50 = 18 nM for wild type 5-HT4 receptors). Binds to receptors in a mechanism different to that of the endogenous ligand 5-HT.

Specifications

Chemical Name or Material 2,3-Dihydro-N-[(3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]-3-(1-methylethyl)-2-oxo-1H-benzimidazole-1-carboxamide hydrochloride
CAS 134296-40-5
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C19H26N4O2.HCl
Formula Weight Observed MW: 378.9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.