Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIN1 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146170DL594
Description
BIN1 Polyclonal antibody specifically detects BIN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
BIN1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
AMPH2, Amphiphysin II, Amphiphysin-like protein, AMPHLDKFZp547F068, box dependant MYC interacting protein 1, bridging integrator 1Box-dependent myc-interacting protein 1, MGC10367, myc box-dependent-interacting protein 1, SH3P9 | |
The immunogen for this product maps to a region between residue 543 and 593 of human Bridging Integrator 1 using the numbering given in entry NP_647593.1 (GeneID 274). | |
0.1 mL | |
Cell Cycle and Replication, Tumor Suppressors | |
274 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction