Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BIN1 Monoclonal Antibody (3B6A4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531651
Description
MA5-31651 has been tested in indirect ELISA.
This gene encodes several isoforms of a nucleocytoplasmic adaptor protein, one of which was initially identified as a MYC-interacting protein with features of a tumor suppressor. Isoforms that are expressed in the central nervous system may be involved in synaptic vesicle endocytosis and may interact with dynamin, synaptojanin, endophilin, and clathrin. Isoforms that are expressed in muscle and ubiquitously expressed isoforms localize to the cytoplasm and nucleus and activate a caspase-independent apoptotic process. Studies in mouse suggest that this gene plays an important role in cardiac muscle development. Alternate splicing of the gene results in ten transcript variants encoding different isoforms. Aberrant splice variants expressed in tumor cell lines have also been described.
Specifications
BIN1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O00499, O08539 | |
BIN1 | |
Purified recombinant fragment of human BIN1 (AA: 189-398) expressed in E. coli. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Western Blot | |
3B6A4 | |
Unconjugated | |
BIN1 | |
ALP-1; AMPH 2; Amph2; amphiphysin 2; amphiphysin II; amphiphysin IIamph2; amphiphysin-like; amphiphysin-like protein; AMPHL; Bin1; box dependant MYC interacting protein 1; box-dependent myc-interacting protein 1; BRAMP-2; bridging integrator 1; DKFZp547F068; MGC10367; MGC105358; myc box dependent interacting protein 1; myc box-dependent-interacting protein 1; SH3 domain-containing protein 9; SH3P9; similar to bridging integrator 1 isoform 9 | |
Mouse | |
Protein G | |
RUO | |
274, 30948 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction